Skip to Content
Merck
All Photos(3)

Key Documents

AV33376

Sigma-Aldrich

Anti-STAT4 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Signal transducer and activator of transcription 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

86 kDa

species reactivity

human, rabbit, horse, dog, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT4(6775)

Immunogen

Synthetic peptide directed towards the N terminal region of human STAT4

Biochem/physiol Actions

STAT4 is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is essential for mediating responses to IL12 in lymphocytes, and regulating the differentiation of T helper cells.

Sequence

Synthetic peptide located within the following region: IGGPLHNGLDQLQNCFTLLAESLFQLRRQLEKLEEQSTKMTYEGDPIPMQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoying Zhou et al.
Helicobacter, 19(3), 174-181 (2014-03-19)
Recent studies found that gastric cancer patients with Helicobacter pylori infection had a better response to chemotherapy and had an improved overall prognosis compared with those without. However, the underlying mechanism remains unknown. Quantitative real-time PCR (qRT-PCR) was utilized to
Lei Lv et al.
Journal of molecular and cellular cardiology, 74, 284-294 (2014-06-17)
Signal transducer and activator of transcription 4 (STAT4) has been associated with susceptibility to autoimmune diseases. Intriguingly, we previously reported that STAT4 might play a critical role in vascular smooth muscle cell (VSMC) proliferation. The present study therefore investigated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service