Skip to Content
Merck
All Photos(1)

Key Documents

278947

Sigma-Aldrich

Bismuth(III) carbonate basic

Synonym(s):

Bismuth subcarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(BiO)2CO3
CAS Number:
Molecular Weight:
509.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

79.1-84.8% Bi basis (EDTA titration)

form

powder

reaction suitability

reagent type: catalyst
core: bismuth

density

6.86 g/mL at 25 °C (lit.)

SMILES string

O=[Bi]OC(=O)O[Bi]=O

InChI

1S/CH2O3.2Bi.2O/c2-1(3)4;;;;/h(H2,2,3,4);;;;/q;2*+1;;/p-2

InChI key

MGLUJXPJRXTKJM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Serena et al.
Advances in skin & wound care, 20(9 Pt 1), 485-492 (2007-09-01)
The human forearm biopsy model can evaluate the effect of novel agents on acute wounds. Bismuth subgallate/borneol (Suile) is a new product cleared by the Food and Drug Administration for treating partial-thickness wounds. Anecdotal reports suggest that Suile may be
[Experimental studies on altering the properties of the prosthetic plastic, Kallocryl B, by adding an x-ray contrast medium].
R Deutschmann et al.
Stomatologie der DDR, 30(7), 475-481 (1980-07-01)
Fan Dong et al.
Dalton transactions (Cambridge, England : 2003), 41(27), 8270-8284 (2012-05-25)
The synthesis and self-assembly of hierarchical architectures from nanoscale building blocks with unique morphology, orientation and dimension have opened up new opportunities to enhance their functional performances and remain a great challenge. This work represents tunable synthesis of various types
Comparison of plasma bismuth levels after oral dosing with basic bismuth carbonate or tripotassium dicitrato bismuthate.
Madaus S, et al.
Alimentary Pharmacology & Therapeutics, 6(2), 241-249 (1992)
Basic metal carbonate supported gold nanoparticles: enhanced performance in aerobic alcohol oxidation.
Yang J, et al.
Green Chemistry, 11(3), 322-325 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service