Skip to Content
Merck
All Photos(1)

Key Documents

61750

Sigma-Aldrich

Lawesson reagent

purum, ≥98.0% (S)

Synonym(s):

2,4-Bis(4-methoxyphenyl)-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-Bis-(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 4-Methoxyphenylthiophosphoric cyclic di(thioanhydride), LR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O2P2S4
CAS Number:
Molecular Weight:
404.47
Beilstein:
1024888
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

purum

Assay

≥98.0% (S)

mp

228-230 °C (lit.)
228-230 °C

SMILES string

COc1ccc(cc1)P2(=S)SP(=S)(S2)c3ccc(OC)cc3

InChI

1S/C14H14O2P2S4/c1-15-11-3-7-13(8-4-11)17(19)21-18(20,22-17)14-9-5-12(16-2)6-10-14/h3-10H,1-2H3

InChI key

CFHGBZLNZZVTAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

The most efficient thiation reagent known till now, used for the thiation of carboxamides, ketones, lactones and imides, lactames, 3-oxocarboxylic derivatives (amides, esters and nitriles) and thio-S-esters; Selective thionation of oligopeptides; Reviews about LR

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I. Thomsen et al.
Organic Syntheses, 62, 158-158 (1984)
M.P. Cava et al.
Tetrahedron, 41, 5061-5061 (1985)
O.E. Jensen et al.
Tetrahedron, 41, 5595-5595 (1985)
R.A. Cherkasov et al.
Tetrahedron, 41, 2567-2567 (1985)
K. Clausen et al.
Journal of the Chemical Society. Perkin Transactions 1, 785-785 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service