Skip to Content
Merck
All Photos(1)

Key Documents

13042

Sigma-Aldrich

N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride

≥99.0% (TLC)

Synonym(s):

N-Benzoyl-L-phenylalanyl-L-valyl-L-arginine-4-nitroanilide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40N8O6 · HCl
CAS Number:
Molecular Weight:
681.18
Beilstein:
3027332
MDL number:
UNSPSC Code:
12352204

Assay

≥99.0% (TLC)

impurities

≤5% water

solubility

methanol: 10 mg/mL, clear, light yellow

SMILES string

Cl.CC(C)[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)c2ccccc2)C(=O)N[C@@H](CCCNC(N)=N)C(=O)Nc3ccc(cc3)[N+]([O-])=O

InChI

1S/C33H40N8O6.ClH/c1-21(2)28(40-31(44)27(20-22-10-5-3-6-11-22)39-29(42)23-12-7-4-8-13-23)32(45)38-26(14-9-19-36-33(34)35)30(43)37-24-15-17-25(18-16-24)41(46)47;/h3-8,10-13,15-18,21,26-28H,9,14,19-20H2,1-2H3,(H,37,43)(H,38,45)(H,39,42)(H,40,44)(H4,34,35,36);1H/t26-,27-,28-;/m0./s1

InChI key

PYVSMZDQQUZGPF-JAQKLANPSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Substrate for thrombin, trypsin and reptilase

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assay of coagulation proteases using peptide chromogenic and fluorogenic substrates.
R Lottenberg et al.
Methods in enzymology, 80 Pt C, 341-361 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service