Skip to Content
Merck
All Photos(3)

Documents

PD2004700

Millipore

Petri dish

47mm petri dish, aseptically molded for microbial application.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122101
eCl@ss:
32030209
NACRES:
NB.14

material

clear device
polystyrene

Quality Level

sterility

sterile

packaging

plate of 150 ×

technique(s)

sterile filtration: suitable

diam.

47 mm

input

sample type food(s)
sample type soft drink
sample type bottled water
sample type beer
sample type beverage(s)
sample type dairy
sample type wine
sample type cider

application(s)

bioburden testing
food and beverages

shipped in

ambient

General description

Petri dish is a single use disposable plastic unit to be used in culturing microorganisms. They are 47mm Petri dishes are aseptically injection molded plates for use with solid agar medium

Application

Petri dish is a single use disposable plastic unit for use with molten agar and to be used in microbiological analysis of Beer, Bottled Water, Cider, Sport Drink, Wine etc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service