Skip to Content
Merck
All Photos(1)

Key Documents

MABC76

Sigma-Aldrich

Anti-Stabilin-2 Antibody, clone mAb-174

clone mAb-174, from mouse

Synonym(s):

Stabilin-2, Fasciclin, EGF-like laminin-type EGF-like and link domain-containing scavenger receptor 2, FAS1 EGF-like and X-link domain-containing adhesion molecule 2, FEEL-2, Hyaluronan receptor for endocytosis, 175 kDa stabilin-2, 175 kDa hyaluronan rec

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

mAb-174, monoclonal

species reactivity

rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Stabilin-2 (Stab2; FEEL-2; and HARE) is a phosphatidylserine receptor that is localized to hepatic sinusoidal endothelial cells. It is a large protein containing a linker domain, two laminin EGF-like domains, four FAS1 domains, and six EGF-like domains, enabling stabilin-2 to perform multiple functions. Stabilin-2 is primarily studied for its role as a scavenger receptor that is involved in the clearance of metabolic waste products, which may include hyaluronic acid, heparin, advanced glycation end-products, acetylated LDL, and chondroitin sulfates. Previous studies have also suggested that stabilin-2 may play a role in cell adhesion processes.

Immunogen

Partially purified rat Stabilin-2 from the HA binding region.

Application

Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in spleen, liver, and lymph node tissues. (Weigel, P., et al. (2003). Journal of Biological Chemistry. 278(11):9808–9812.)
This Anti-Stabilin-2 Antibody, clone mAb-174 is validated for use in Western Blotting, IHC for the detection of Stabilin-2.

Quality

Evaluated by Western Blot in rat spleen tissue lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected Stabilin-2 on 10 µg of rat spleen tissue lysate.

Target description

~190 kDa observed. The calculated molecular weight of this protein is 155 kDa, but can be observed at ~190 kDa due to high glycosylation. An uncharacterized band appears at ~20 kDa in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service