Skip to Content
Merck
All Photos(1)

Documents

8.56070

Sigma-Aldrich

H-Gln(Trt)-Sulfamylbutyryl NovaSyn® TG resin

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

NovaSyn® TG
Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

Pre-loaded resin for the synthesis of C-terminal safety-catch resin for synthesis of C-terminally modified glutamine peptides by Fmoc SPPS. Linker is activated to cleavage by nucleophiles by methylation using TMS-diazomethane.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Linkage

Replaces: 04-12-3717

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.10 - 0.30 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The base resin is amino PEG-PS-polymer (90 µm).

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service