Skip to Content
Merck
All Photos(1)

Documents

8.43849

Sigma-Aldrich

Potassium tert-butylate

(~ 20% solution in tetrahydrofuran) for synthesis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.22

Quality Level

form

liquid

transition temp

flash point -21.5 °C (THF)

density

0.914 g/cm3 at 20 °C

storage temp.

2-8°C

Analysis Note

Assay (acidimetric): 19.0 - 21.0 %
Density (d 20 °C/ 4 °C): 0.912 - 0.917

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 2 - Skin Corr. 1A - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

-6.7 °F - (THF)

Flash Point(C)

-21.5 °C - (THF)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service