Skip to Content
Merck
All Photos(1)

Documents

MANH95385420

Manchester Organics

Potassium tetrakis(pentafluorophenyl)borate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
718.14
MDL number:

Iupac Name

potassium tetrakis(2,3,4,5,6-pentafluorophenyl)borate(1-)

Inchi Code

1S/C24BF20.K/c26-5-1(6(27)14(35)21(42)13(5)34)25(2-7(28)15(36)22(43)16(37)8(2)29,3-9(30)17(38)23(44)18(39)10(3)31)4-11(32)19(40)24(45)20(41)12(4)33;/q-1;+1

InChI key

GYBHRIJOPWTIKA-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/J64639

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service