Skip to Content
Merck
All Photos(1)

Documents

APO455827424

Apollo Scientific Ltd

3-(1H-Imidazol-1-yl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
140.14
MDL number:

bp

147-149

Iupac Name

3-(1H-imidazol-1-yl)propanoic acid

Inchi Code

1S/C6H8N2O2/c9-6(10)1-3-8-4-2-7-5-8/h2,4-5H,1,3H2,(H,9,10)

InChI key

VSFNAZLYGOOSEY-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/3-1h-imidazol-1-ylpropanoic-acid

Boiling point

147-149

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-1h-imidazol-1-ylpropanoic-acid


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service