W302503
Sodium benzoate
≥99%, FCC, FG
Synonym(s):
Benzoic acid sodium salt
About This Item
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Halal
Kosher
reg. compliance
EU Regulation 1333/2008 & 178/2002
FCC
FDA 21 CFR 150.141
FDA 21 CFR 150.161
FDA 21 CFR 166.110
FDA 21 CFR 181.23
FDA 21 CFR 184.1733
Assay
≥99%
mp
>300 °C (lit.)
solubility
water: soluble 0.5 g/mL
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
odorless
SMILES string
[Na+].[O-]C(=O)c1ccccc1
InChI
1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1
InChI key
WXMKPNITSTVMEF-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service