Skip to Content
Merck
All Photos(2)

Key Documents

ALD00008

Sigma-Aldrich

2-((N-Succinimidyl)oxy)-2-methylpropanoic acid

95% (GC)

Synonym(s):

Baran Aminating Reagent Precursor

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R 1 212,26

R 1 212,26

List PriceR 1 237,00

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R 1 212,26

About This Item

Empirical Formula (Hill Notation):
C8H11NO5
Molecular Weight:
201.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

R 1 212,26

List PriceR 1 237,00

Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

95% (GC)

form

solid

storage temp.

2-8°C

SMILES string

OC(C(C)(C)ON1C(CCC1=O)=O)=O

InChI

1S/C8H11NO5/c1-8(2,7(12)13)14-9-5(10)3-4-6(9)11/h3-4H2,1-2H3,(H,12,13)

InChI key

DHXJNWNKFUOFOR-UHFFFAOYSA-N

Application

Immediate precursor to a perester-containing imidating reagent; which performs a net amination of aromatic and heteroaromatic substrates.

Learn More at the Professor and Product Portal of Professor Phil S. Baran

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service