Skip to Content
Merck
All Photos(1)

Key Documents

772291

Sigma-Aldrich

4-Methoxyphenyl 2,3,4,6-tetra-O-acetyl-α-D-mannopyranoside

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O11
CAS Number:
Molecular Weight:
454.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

optical activity

[α]22/D +70.0°, c = 1% in chloroform

mp

100-104 °C

storage temp.

2-8°C

SMILES string

COc1ccc(O[C@H]2O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@@H]2OC(C)=O)cc1

InChI

1S/C21H26O11/c1-11(22)27-10-17-18(28-12(2)23)19(29-13(3)24)20(30-14(4)25)21(32-17)31-16-8-6-15(26-5)7-9-16/h6-9,17-21H,10H2,1-5H3/t17-,18-,19+,20+,21+/m1/s1

InChI key

RPHXBVOPPUTUES-MJCUULBUSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service