Skip to Content
Merck
All Photos(1)

Key Documents

49581

Sigma-Aldrich

Hexadecyl(2-hydroxyethyl)dimethylammonium dihydrogen phosphate solution

~30% in H2O

Synonym(s):

Cetyldimethyl(2-hydroxyethyl)ammonium dihydrogen phosphate, Dimethylhexadecyl(2-hydroxyethyl)ammonium dihydrogen phosphate, Luviquat mono CP AT1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(CH3)2(CH2CH2OH)[OPO(OH)2]
CAS Number:
Molecular Weight:
411.56
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

description

cationic

mol wt

411.56 g/mol

concentration

~30% in H2O

refractive index

n20/D 1.381

application(s)

detection

SMILES string

OP(O)([O-])=O.CCCCCCCCCCCCCCCC[N+](C)(C)CCO

InChI

1S/C20H44NO.H3O4P/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-21(2,3)19-20-22;1-5(2,3)4/h22H,4-20H2,1-3H3;(H3,1,2,3,4)/q+1;/p-1

InChI key

SGZVXLFVBKDMJH-UHFFFAOYSA-M

Legal Information

Luviquat is a trademark of BASF SE

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamil Peckh et al.
Materials (Basel, Switzerland), 13(20) (2020-10-18)
Studies on the oxidation of α-olefins via the two-stage method are presented. The new method consisted of oxidizing C30+ α-olefins with hydrogen peroxide (2 equiv.) and subsequent oxidation with oxygen. Products with high acid numbers (29-82 mgKOH/g) and saponification numbers
Davide Albani et al.
Nature communications, 9(1), 2634-2634 (2018-07-08)
Ensemble control has been intensively pursued for decades to identify sustainable alternatives to the Lindlar catalyst (PdPb/CaCO3) applied for the partial hydrogenation of alkynes in industrial organic synthesis. Although the geometric and electronic requirements are known, a literature survey illustrates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service