Skip to Content
Merck
All Photos(1)

Key Documents

473812

Sigma-Aldrich

1,1,2,2-Tetrafluoro-3-iodopropane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2CF2CHF2
CAS Number:
Molecular Weight:
241.95
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.414 (lit.)

bp

99 °C (lit.)

density

2.117 g/mL at 25 °C (lit.)

SMILES string

FC(F)C(F)(F)CI

InChI

1S/C3H3F4I/c4-2(5)3(6,7)1-8/h2H,1H2

InChI key

VOUMUNAZCCDSHT-UHFFFAOYSA-N

General description

1,1,2,2-Tetrafluoro-3-iodopropane is a fluorinated building block. Rotational transitions and complete iodine nuclear quadrupole coupling tensor of its trans-trans and gauche-gauche conformers have been determined.[1]

Application

1,1,2,2-Tetrafluoro-3-iodopropane may be used in the iodine transfer polymerization (ITP) of vinylidene fluoride (VDF).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodine transfer polymerization (ITP) of vinylidene fluoride (VDF). Influence of the defect of VDF chaining on the control of ITP.
Boyer C, et al.
Macromolecules, 38(25), 10353-10362 (2005)
Chirped pulse Fourier transform microwave spectroscopy of 1, 1, 2, 2-tetrafluoro-3-iodopropane.
Grubbs GS, et al.
Molecular Physics, 107(21), 2221-2225 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service