Skip to Content
Merck
All Photos(1)

Key Documents

440035

Sigma-Aldrich

Ethyl 4-(2-chloroacetamido)benzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CONHC6H4CO2C2H5
CAS Number:
Molecular Weight:
241.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

110-114 °C (lit.)

SMILES string

CCOC(=O)c1ccc(NC(=O)CCl)cc1

InChI

1S/C11H12ClNO3/c1-2-16-11(15)8-3-5-9(6-4-8)13-10(14)7-12/h3-6H,2,7H2,1H3,(H,13,14)

InChI key

ZVRJEYAQESBSSH-UHFFFAOYSA-N

General description

Ethyl 4-(2-chloroacetamido)benzoate can be synthesized starting from benzocaine. It undergoes heterocyclization in the presence of ammonium thiocyanate in refluxing ethanol to afford ethyl-4-(4-oxothiazolidin-2-ylideneamino)benzoate, via intramolecular cyclization and the Dimroth-like rearrangements.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haider Behbehani et al.
Molecules (Basel, Switzerland), 17(6), 6362-6385 (2012-05-29)
The 4-thiazolidinones 3a-d were used as a key intermediates for the synthesis of 2-arylimino-5-arylidene-4-thiazolidinones derivatives 7a–p via nucleophilic addition reactions with the arylidene malononitrile. Moreover the 4-thiazolidinones 3a and 3c condensed with the DMF-DMA to form the corresponding enamines 8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service