Skip to Content
Merck
All Photos(1)

Documents

436003

Sigma-Aldrich

Silver(I) hydrogenfluoride

97%

Synonym(s):

Silver(I) bifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgHF2
CAS Number:
Molecular Weight:
146.87
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

97%

form

crystalline

SMILES string

F.F[Ag]

InChI

1S/Ag.2FH/h;2*1H/q+1;;/p-1

InChI key

BABIQFTUTNDWMB-UHFFFAOYSA-M

Application

Facilitates the photochemical conversion of carboxylic acids to alkyl fluorides.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lai, C. et al.
The Journal of Organic Chemistry, 58, 1393-1393 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service