Skip to Content
Merck
All Photos(1)

Documents

210803

Sigma-Aldrich

Acid Violet 5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20N4Na2O10S3
CAS Number:
Molecular Weight:
678.62
Colour Index Number:
18125
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

mp

240 °C (dec.) (lit.)

λmax

527 nm

SMILES string

[Na+].[Na+].CC(=O)Nc1ccc(cc1)\N=N\c2c(O)c3c(NS(=O)(=O)c4ccc(C)cc4)cc(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C25H22N4O10S3.2Na/c1-14-3-9-19(10-4-14)40(32,33)29-21-13-20(41(34,35)36)11-16-12-22(42(37,38)39)24(25(31)23(16)21)28-27-18-7-5-17(6-8-18)26-15(2)30;;/h3-13,29,31H,1-2H3,(H,26,30)(H,34,35,36)(H,37,38,39);;/q;2*+1/p-2/b28-27+;;

InChI key

RHEGITQOJDSUSE-JZYARHMISA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service