Documents
23238
Supelco
Screw cap
black phenolic hole cap (with polyseal liner), thread for 20-400, pkg of 144 ea
About This Item
UNSPSC Code:
41122001
material
black phenolic hole cap (with polyseal liner)
packaging
pkg of 144 ea
fitting
thread for 20-400
compatibility
for use with 30 mL (32 × 80 mm) ~ For Bottle (Volume and Dimensions): 60 mL (38 × 95 mm)
General description
Screw caps are available as hole caps for use with sealing septa. Black phenolic hole cap (with polyseal liner, thread 20-400) can withstand a temperature of 149°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)