Skip to Content
Merck
All Photos(1)

Key Documents

870747P

Avanti

24:0(d4) Coenzyme A

Avanti Research - A Croda Brand, powder

Synonym(s):

Lignoceroyl(d4) coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H87D4N10O17P3S
CAS Number:
Molecular Weight:
1173.27
UNSPSC Code:
12352209
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (870747P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCC([2H])(C(CCCCCCCCCCC)([2H])[2H])[2H])=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

General description

Lignoceroyl(d4) coenzyme A (ammonium salt) (24:0(d4) Coenzyme A) is a deuterated derivative of 24:0 Coenzyme A.

Packaging

5 mL Amber Glass Screw Cap Vial (870747P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service