Skip to Content
Merck
All Photos(1)

Key Documents

933538

Sigma-Aldrich

AFM Disc Gripper

disc diam. 12 mm

Synonym(s):

Disc Grippers for AFM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

disc diam.

12 mm

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

AFM Disc Gripper, 12 mm, can be used to pick up Atomic Force Microscope (AFM)/ scanning tunneling microscope (STM) discs easily from a flat surface. Features a protective rubber-coated tip.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service