Skip to Content
Merck
All Photos(2)

Key Documents

101990

Sigma-Aldrich

4,4′-Dibromooctafluorobiphenyl

99%

Synonym(s):

1-Bromo-4-(4-bromo-2,3,5,6-tetrafluorophenyl)-2,3,5,6-tetrafluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6F4C6F4Br
CAS Number:
Molecular Weight:
455.92
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

113-115 °C (lit.)

functional group

bromo
fluoro

SMILES string

Fc1c(F)c(c(F)c(F)c1Br)-c2c(F)c(F)c(Br)c(F)c2F

InChI

1S/C12Br2F8/c13-3-9(19)5(15)1(6(16)10(3)20)2-7(17)11(21)4(14)12(22)8(2)18

InChI key

YXLMNFVUNLCJJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Dibromooctafluorobiphenyl undergoes lithiation with n-butyllithium to yield 4,4′-dilithiooctafluorobiphenyl and 4-lithio-4′-bromooctafluorobiphenyl.

Application

4,4′-Dibromooctafluorobiphenyl was used as internal standard to determine brominated flame retardants in sediment samples using on-line combination of pressurised hot water extraction and liquid chromatography-gas chromatography.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Reactions and thermal decomposition of organolithium compounds derived from 4, 4'-dibromooctafluorobiphenyl.
Contigiani RA, et al.
Journal of Organometallic Chemistry, 32(1), 7-15 (1971)
Huizhen Li et al.
Chemosphere, 144, 2427-2435 (2015-11-30)
Commercial mosquito repellents (MRs) are generally applied as mosquito coils, electric vaporizers (liquid and solid) or aerosol spray, with pyrethroids often being the active ingredients. Four types of MRs were applied individually in a 13-m(2) bedroom to study the occurrence
Kati Kuosmanen et al.
Journal of chromatography. A, 943(1), 113-122 (2002-02-01)
Pressurised hot water extraction (PHWE) was coupled on-line with liquid chromatography-gas chromatography (LC-GC) to determine brominated flame retardants in sediment samples. After extraction with pressurised hot water the analytes were adsorbed in a solid-phase trap. The trap was dried with
Shunhui Wang et al.
Journal of hazardous materials, 393, 122420-122420 (2020-03-07)
Fipronil and its degradates have been detected ubiquitously in aquatic environment worldwide, yet little is known about its bioaccumulation potential. The goal of the present study was to measure bioconcentration factor (BCF) of sediment-associated fipronil in a benthic invertebrate, Lumbriculus
Yuanyuan Pei et al.
Marine pollution bulletin, 124(2), 1048-1054 (2016-11-09)
The occurrence of polycyclic aromatic hydrocarbons (PAHs) and polychlorinated biphenyl (PCBs) in urban atmosphere in Guangzhou, China were assessed using homing pigeons as a biomonitor. Contaminant concentrations in lung were significantly higher than those in liver and fat, indicating chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service