Principaux documents
K5170-U
Omnifit® EZ Benchmark Replacement Glass Columns
I.D. × L 35 mm × 150 mm
About This Item
Code UNSPSC :
23151816
Nomenclature NACRES :
SB.52
Fabricant/nom de marque
Omnifit 006ERG-35-15
Niveau de qualité
Technique(s)
LPLC: suitable
D.I. × L
35 mm × 150 mm
Description générale
Replacement glass columns are available in a wide range of bore sizes and lengths. This not only enables you to replace a column should breakage occur, but allows you to obtain several different length columns to use with endpieces you already have for that bore size. The modular nature of our column ranges means that for the ultimate flexibility you can mix and match column sizes with different endpiece types to your specific requirements.
Informations légales
Omnifit is a registered trademark of Diba Industries