Principaux documents
21038
Supelco
Filtre en quartz, diamètre 32mm
(d)
About This Item
Code UNSPSC :
12190000
Conditionnement
pkg of 10 ea
Technique(s)
active air sampling: suitable
Application(s)
air monitoring
environmental
industrial hygiene
Description générale
Quartz Filter along with filter cartridge assembly enables simultaneous sampling of both the gas and particulate fractions.
Andressa Amado Martin et al.
International journal of biological macromolecules, 144, 527-535 (2019-12-21)
Hyaluronic acid (HA) and galactomannans (such as guar gum - GG and locust bean gum - LBG) are biomacromolecules with various applications due to their physicochemical characteristics in solution, especially their ability to form films or hydrogels in some conditions.