Principaux documents
OH30456410
Ohaus® Navigator® Portable Balance
Model NV222, weighing capacity 220 g, resolution: 0.01 g, AC/DC input 110 V, US 3-pin plug (North America)
About This Item
Code UNSPSC :
41111502
Matériaux
metal weighing plate
Caractéristiques
CSA approved
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30456410
Plage de mesure de la capacité de pesée
220 g, resolution: 0.01 g
Diam. de la nacelle de pesée
14.5 cm
Description générale
OHAUS raises the bar in price-performance ratio with the newly-designed Navigator series which is perfect for general weighing applications in laboratory, industrial and education settings. Ruggedly constructed and multifunctional, the Navigator′s a powerful balance that can handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity, and easy operation.
Application
Weighing, Parts Counting, Percent Weighing, Checkweighing
Caractéristiques et avantages
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Informations légales
Navigator is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.