CLS440141
Corning® temperature probe support clamp
Synonyme(s) :
lab equipment accessory
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.76
Produits recommandés
Fabricant/nom de marque
Corning 440141
L × D.E.
16.5 mm × 11 mm
Description générale
Corning offers a 7/16 in. (1.1 cm) diameter by 6 1/2 in. (16.5 cm) support clamp. Support has two openings, 1/3 in. (9 mm) and 0.15 in. (3.75 mm), for holding probes. It is designed to be used with a boss head clamp
Informations légales
Corning is a registered trademark of Corning, Inc.