Principaux documents
Z767328
Microflex NeoPro® EC powder-free chloroprene gloves
size S
Synonyme(s) :
disposable glove
About This Item
Code UNSPSC :
46181504
Matériaux
(chloroprene gloves)
green
Taille
S
Description
ambidexterous
textured fingers
Caractéristiques
CE compliant
disposable
latex free
powder-free
Conditionnement
pack of 50 ea (boxes of 50)
Fabricant/nom de marque
Microflex NEC-288-S
Longueur
11.8 in. , cuff
Épaisseur
6.3 mil , palm
8.3 mil , fingers
Catégories apparentées
Description générale
Featureing an extended cuff, NeoPro® EC is the longest and thickest non-latex glove produced by Microflex. It uses a polychloroprene formulation to provide a secure grip along with exceptional protection from biological and chemical hazards inwet or dry conditions.
Informations légales
NeoPro is a registered trademark of CIT Healthcare LLC