Principaux documents
Z556734
Sigma-Aldrich
Aldrich® Benchclamp 2-prong
dual adjustment, size M
Synonyme(s) :
bench clamps
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
Taille
M
L × diam.
127 mm × 11 mm , arm
Capacité de la mâchoire
0-75 mm
Largeur de la fixation
23 mm
Description générale
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC