Z409480
Weighing funnel
size M, polypropylene
Synonyme(s) :
Weighing Funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.71
Produits recommandés
Matériaux
polypropylene
Taille
M
Conditionnement
pkg of 50 ea
Description générale
- Sit flat on balance - No rolling, no spilling!
- Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
- Residual sample can be rinsed to guarantee 100% transfer
- Suitable for safe handling of hazardous materials
- Available in five different sizes: extra small, small, medium, large, and extra large