Key Documents
Z126586
Sigma-Aldrich
Recessed septa mixed set
set of 1
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.35
Matériaux
rubber septa (silicone)
Conditionnement
set of 1
Description générale
Contains ten of each size for the following septa, with carrying box:
Fitting Neck I.D.
8.7 mm
10.7 mm
15.0 mm
17.0 mm
23.0 mm