Principaux documents
Z118028
Luer-to-Tubing Connector
male Luer lock to hose end, for tubing i.d., 1/16 ‑ 3/32 in., nickel plated
Synonyme(s) :
environmental, veterinary
About This Item
Code UNSPSC :
40183110
Nomenclature NACRES :
NB.03
Matériaux
nickel plated
Caractéristiques
for tubing i.d. 1/16 ‑ 3/32 in.
Fabricant/nom de marque
Cadence Science 6157
Catégories apparentées
Description générale
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating, easy to clean and resistant to corrosion.
Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.