Principaux documents
TEM-CF200CU
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
size 200 mesh, box of 25
Synonyme(s) :
CARBON FILM 200 MESH CU 25/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 25
Taille
200 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Copper grid, 200 mesh.