Principaux documents
SYNF642460
Synthware pressure equalizing funnel with high vacuum valve
60 mL, joint: ST/NS 24/40, valve size 0-4 mm
Synonyme(s) :
Synthware F642460, addition funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F642460
Capacité
60 mL
60 mL
Description générale
With PTFE extended tip valve for semi-needle valve adjustment.
Informations légales
Synthware is a trademark of Kemtech America Inc.