Principaux documents
SYNF641960
Synthware pressure equalizing funnel with high vacuum valve
60 mL, joint: ST/NS 19/22, valve size 0-4 mm
Synonyme(s) :
Synthware F641960, addition funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F641960
Capacité
60 mL
60 mL
Catégories apparentées
Description générale
With PTFE extended tip valve for semi-needle valve adjustment.
Informations légales
Synthware is a trademark of Kemtech America Inc.