Principaux documents
SYNF341110
Synthware two neck pear shaped flask
10 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonyme(s) :
Synthware F341110, 2-neck pear shape flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
pear-shaped
two-neck (angled side neck)
Joint
center joint: ST/NS14/20
side joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F341110
Capacité
10 mL
10 mL
Description générale
Pear shape with joints. Heavy wall design.
Informations légales
Synthware is a trademark of Kemtech America Inc.