Principaux documents
CLS808050
Pyrex® centrifuge tubes with beaded rim and white enamel graduations
capacity 50 mL
Synonyme(s) :
Corning® 8080
About This Item
Code UNSPSC :
41121703
Nomenclature NACRES :
NB.43
Matériaux
conical bottom tube
Stérilité
non-sterile
Conditionnement
case of 12
pack of 6
Fabricant/nom de marque
Corning 8080-50
D.E. × L
28 mm × 135 mm
Capacité
50 mL
Description générale
Formed from borosilicate glass. Stress that glass can withstand is greatly decreased by surface flaws such as scratches, chips, and cracks. Always use with correct sleeve or holder.
Autres remarques
15 mL sizes have 0.1 mL graduations upward from tip; 50 mL size has 0.5 mL graduations upward from tip to 10 mL and 1 mL graduations from 10 to 50 mL.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.