Documents
CLS4965B500
Pyrex® three-neck round-bottom flask
capacity 500 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22
Synonyme(s) :
Pyrex® round-bottom three neck angle type distilling flask, bottom flask, flask round, flask round bottom
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom flask
three-neck (Vertical)
Joint
center joint: ST/NS19/22
side joint: ST/NS19/22
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper joint
Conditionnement
case of 1
Fabricant/nom de marque
Corning 4965B-500
Capacité
500 mL
Taille du col
(3)
Informations légales
PYREX is a registered trademark of Corning, Inc.