Principaux documents
BR749321
BRAND® micro haematocrit capillary
non-hep.
Synonyme(s) :
Haematocrit capillaries
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.22
Matériaux
blue
Caractéristiques
DIN ISO 12772 and BS 4316-68, To Contain
Conditionnement
pack of 1000 ea
Fabricant/nom de marque
BRAND 749321
Catégories apparentées
Description générale
Compliant with DIN ISO 12772 and BS 4316-68 standards, these tubes feature a color code for easy identification and use in hematocrit centrifuges. The heparinized tubes are treated with sodium heparinate on the internal surface and are marked with a red color code, making them suitable for capillary blood samples. In contrast, the non-heparinized tubes are designated with a blue color code, specifically designed for use with heparinized venous blood.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Guang Yang et al.
Journal of cellular and molecular medicine, 19(10), 2413-2422 (2015-07-16)
Previous work from both our lab and others have indicated that exposure to 50 Hz magnetic fields (ELF-MF) was able to modify ion channel functions. However, very few studies have investigated the effects of MF on γ-aminobutyric acid (GABA) type