Principaux documents
BAH131781220
SP Bel-Art® utility bags
width × H × thickness 12 in. (305 mm) × 20 in. (508 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Synonyme(s) :
clear bag, reusable bag, sample bag
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Matériaux
clear
Conditionnement
pack of 100 ea
Fabricant/nom de marque
Bel-Art H13178-1220
width × H × thickness
12 in. (305 mm) × 20 in. (508 mm) × 4 mil (0.1 mm)
Description générale
Reusable, transparent bags are ideal for shipping or storage and are especially useful as sample bags.
Caractéristiques et avantages
  • FDA grade polyethylene, safe for food, drug and cosmetic use
  • Latex free
  • Not Autoclavable
  • H13176 lightweight bags are 0.038 mm (1.5 mil) thick, H13178 heavyweight bags are 0.1016 mm (4 mil) thick
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP