Merck
Rh-catalyzed linear hydroformylation of styrene.
Dalton transactions (Cambridge, England : 2003) (2012-10-30)
Evert Boymans, Michèle Janssen, Christian Müller, Martin Lutz, Dieter Vogt
ABSTRACT
Usually the Rh-catalyzed hydroformylation of styrene predominantly yields the branched, chiral aldehyde. An inversion of regioselectivity can be achieved using strong π-acceptor ligands. Binaphthol-based diphosphite and bis(dipyrrolyl-phosphorodiamidite) ligands were applied in the Rh-catalyzed hydroformylation of styrene. High selectivities up to 83% of 3-phenylpropanal were obtained with 1,1-bi-2-naphthol-based bis(dipyrrolyl-phosphorodiamidite) with virtually no hydrogenation to ethyl benzene. The coordination chemistry of those ligands towards Rh(I) was investigated spectroscopically and structurally.
Sigma-Aldrich
Styrene, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Styrene, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrocinnamaldehyde, technical grade, 90%