Skip to Content
Merck
All Photos(1)

Key Documents

AB15115

Sigma-Aldrich

Anti-NARC-1 Antibody

serum, Chemicon®

Synonym(s):

Neural Apoptosis-Regulated Convertase 1, PCSK9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
mouse
technique(s):
western blot: suitable
citations:

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Pcsk9(100102)

Specificity

Neural apoptosis-regulated convertase 1 [NARC-1, Pcsk9]. By Western blot the antibody recognizes a doublet at ~76 kDa in mouse liver lysate.

Immunogen

Synthetic peptide.

Application

NARC-1 is an antibody targeting the NARC-1 protein, validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Physical form

Rabbit serum.
Serum

Storage and Stability

Maintain at -20degC in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service