Skip to Content
Merck
All Photos(1)

Key Documents

70181-M

Sigma-Aldrich

NovaBlue Singles Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

Competent E. coli Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
biological source:
Escherichia coli
growth mode:
adherent or suspension
morphology:
rod shaped
manufacturer/tradename:
Novagen®

Product Name

NovaBlue Singles Competent Cells - Novagen, NovaBlue Singles Competent Cells are designed for ultimate convenience and reliability in plasmid transformation.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >1.5×108 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype:

endA1 hsdR17 (rK12 mK12+) supE44 thi-1 recA1 gyrA96 relA1 lac F′[proA+B+ lacIqZΔM15::Tn10] (TetR)





This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
NovaBlue Singles Competent Cells are designed for ultimate convenience and reliability in plasmid transformation.
NovaBlue competent cells are for routine molecular cloning applications. NovaBlue is a K-12 strain derivative that offers high transformation efficiency,facilitates plasmid stability, and allows blue/whitescreening with appropriate plasmids.

NovaBlue Singles Competent Cells, like all Novagen Singles Competent Cells, are designed for ultimate convenience and reliability in plasmid transformation. The cells are grown and made chemically competent by an optimized procedure. Cells are provided in 50-l volumes that eliminate the need to aliquot, freeze/thaw, or waste partially used vials, thus saving time and increasing performance. For use, simply thaw, add DNA, incubate five minutes on ice, heat shock for 30 seconds, place on ice for two minutes, add SOC Medium, and plate directly (when selecting for ampicillin resistance) or after incubation at 37°C for 30 minutes (when selecting for kanamycin resistance).

Components

•11 or 22Singles Competent Cells•2 or 4 × 2 mlSOC Medium•10 µlTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

DNA Profile

Genotype: endA1 hsdR17 (rK12 mK12+) supE44 thi-1 recA1 gyrA96 relA1 lac Fʹ[proA+B+ lacIqZΔM15::Tn10] (TetR)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service