Skip to Content
Merck
All Photos(2)

Key Documents

5.34354

Sigma-Aldrich

p300/CBP Inhibitor X, EML425

Synonym(s):

p300/CBP Inhibitor X, EML425, 1,3-dibenzyl-5-(4-hydroxy-2,6-dimethylbenzylidene)pyrimidine-2,4,6(1H,3H,5H)-trione, EML-425, p300/CBP Acetyltransferase Inhibitor, EML425

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H24N2O4
Molecular Weight:
440.49
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable benzylidenbarbituric acid derivative that acts as a potent, selective inhibitor of lysine acetyltransferase p300/CBP paralogues (IC50 = 2.9 µM and 1.1 µM for recombinant p300 and CBP, respectively). The inhibition appears to be reversible and is non-competitive with respect to both acetyl-CoA and a histone H3 peptide. It binds to both the free enzyme and the enzyme-substrate complex with uneven affinities. Does not affect the activity of general control non-derepressible 5 (GCN5) and p300/CBP-associated factor (PCAF). Shown to reduce the acetylation of lysine H4K5 and H3K9 in U937 cells (~100 µM) and induce a cell cycle arrest at the G0/G1 phase with an increase in the percentage of hypodiploid nuclei (~ 100 µM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p300/CBP
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Milite, C., et al. 2015. J. Med. Chem.58, in press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service