GF23140820
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1
foil, 85 x 85mm, 0.1mm thickness, annealed
Synonym(s):
FE080261
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
foil (annealed)
manufacturer/tradename
Goodfellow 231-408-20
size × thickness
85 mm × 85 mm × 0.1 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service