Merck
  • Reactions of indoles with nitrogen dioxide and nitrous acid in an aprotic solvent.
Reactions of indoles with nitrogen dioxide and nitrous acid in an aprotic solvent.
Organic & biomolecular chemistry (2006-10-13)
Paola Astolfi, Maria Panagiotaki, Corrado Rizzoli, Lucedio Greci
ABSTRACT
The reaction of 2-phenyl- and 1-methyl-2-phenylindole with nitrogen dioxide or with nitrous acid (NaNO2-CH3COOH) in benzene leads mainly to the formation of the isonitroso and 3-nitroso indole derivatives, respectively. When reacted with nitrous acid, 1-methyl-2-phenylindole gives also the corresponding azo-bis-indole in good yields. The reaction of indole with nitrogen dioxide leads to 2-(indol-3-yl)-3H-indol-3-one as the main product together with small amounts of 2-(indol-3-yl)-3H-indol-3-oxime; whereas the major product obtained when the same indole is reacted with nitrous acid is represented by 2-(indol-3-yl)-3H-indol-3-oxime. The reaction of 3-alkyl substituted indoles with nitrogen dioxide is rather complex and results in the formation of different nitro indoles, whereas nitrosation is observed when nitrous acid is used. Crystal structures of 2-(indol-3-yl)-3H-indol-3-one and of 4-nitro-N-acetyltryptamine have been determined by X-ray analysis.
Sigma-Aldrich
1-Methyl-2-phenylindole, 99%
Pricing and availability is not currently available.