Merck
  • Prenatal exposure to environmental contaminants and body composition at age 7-9 years.
Prenatal exposure to environmental contaminants and body composition at age 7-9 years.
Environmental research (2014-04-20)
Immle Delvaux, Jolijn Van Cauwenberghe, Elly Den Hond, Greet Schoeters, Eva Govarts, Vera Nelen, Willy Baeyens, Nicolas Van Larebeke, Isabelle Sioen
ABSTRACT
The study aim was to investigate the association between prenatal exposure to endocrine disrupting chemicals (EDCs) and the body composition of 7 to 9 year old Flemish children. The subjects were 114 Flemish children (50% boys) that took part in the first Flemish Environment and Health Study (2002-2006). Cadmium, PCBs, dioxins, p,p'-DDE and HCB were analysed in cord blood/plasma. When the child reached 7-9 years, height, weight, waist circumference and skinfolds were measured. Significant associations between prenatal exposure to EDCs and indicators of body composition were only found in girls. After adjustment for confounders and covariates, a significant negative association was found in girls between prenatal cadmium exposure and weight, BMI and waist circumference (indicator of abdominal fat) and the sum of four skinfolds (indicator of subcutaneous fat). In contrast, a significant positive association (after adjustment for confounders/covariates) was found between prenatal p,p'-DDE exposure and waist circumference as well as waist/height ratio in girls (indicators of abdominal fat). No significant associations were found for prenatal PCBs, dioxins and HCB exposure after adjustment for confounders/covariates. This study suggests a positive association between prenatal p,p'-DDE exposure and indicators of abdominal fat and a negative association between prenatal cadmium exposure and indicators of both abdominal as well as subcutaneous fat in girls between 7 and 9 years old.
Product Description
Supelco
4,4′-DDE, PESTANAL®, analytical standard
Supelco
4,4′-DDMU, PESTANAL®, analytical standard
Sigma-Aldrich
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene, 99%