Skip to Content
Merck
All Photos(1)

Key Documents

35489

Supelco

4,4′-DDMU

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-1,1-bis(4-chlorophenyl)-ethene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl3
CAS Number:
Molecular Weight:
283.58
Beilstein:
1461623
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cl\C=C(\c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C14H9Cl3/c15-9-14(10-1-5-12(16)6-2-10)11-3-7-13(17)8-4-11/h1-9H

InChI key

LNKQQZFLNUVWQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiong An et al.
Huan jing ke xue= Huanjing kexue, 28(12), 2794-2799 (2008-02-23)
Dynamics of DDMS and DDMU in soils with DDT application under rice and ryegrass planting by pot experiment in greenhouse was conducted. Results show that DDE/DDD is slightly increased, and only about 20 ng x g(-1) DDMU or DDMS could
Erica F Pirnie et al.
Chemosphere, 65(9), 1576-1582 (2006-05-09)
Significant concentrations of chlorinated pesticides such as 1,1,1-trichloro-2,2-bis(4-chlorophenyl)ethane (DDT) and its two main transformation products, 1,1-dichloro-2,2-bis(4-chlorophenyl)ethane (DDD) and 1,1-dichloro-2,2-bis(4-chlorophenyl)ethylene (DDE) are still present in soil and sediment systems more than 30 years after DDT use was banned in the United
Panu Rantakokko et al.
Journal of chromatography. A, 1216(6), 897-901 (2008-12-26)
A simple and fast method is presented to be used for example in studies on the relationship between serum levels of persistent organic pollutants and type 2 diabetes mellitus. Method is based on liquid-liquid extraction and gas chromatography coupled with
Delphine Bachelet et al.
Environmental research, 111(6), 861-870 (2011-06-21)
Dichlorodiphenyltrichloroethane (DDT) and polychlorinated biphenyls (PCBs) are persistent organochlorine compounds bioaccumulating in human tissues. Body burden of organochlorines may be influenced by individual characteristics such as age, weight variations, breastfeeding, dietary habits and place of residence. To assess the current
Lea A Cupul-Uicab et al.
Environmental research, 110(6), 595-603 (2010-06-23)
Recent data suggest that prenatal exposure to p,p'-DDE may reduce height and increase body mass index (BMI) in childhood, thus potentially raising the risk of adult health problems. The association between prenatal DDE exposure and growth was evaluated in 788

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service