Skip to Content
Merck
o-Fluoro-DL-phenylalanine ≥98%

o-Fluoro-DL-phenylalanine

o-Fluoro-DL-phenylalanine ≥98%
Linear Formula:
FC6H4CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
183.18
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing