Skip to Content
Merck
All Photos(1)

Key Documents

N11779

Cacodylic acid

analytical standard

Synonym(s):

Dimethylarsinic acid, Dimethylarsonic acid, Hydroxydimethylarsine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2As(O)OH
CAS Number:
Molecular Weight:
138.00
Beilstein:
1736965
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 500 mg

manufacturer/tradename

Chem Service, Inc. PS-51

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C[As](C)(O)=O

InChI

1S/C2H7AsO2/c1-3(2,4)5/h1-2H3,(H,4,5)

InChI key

OGGXGZAMXPVRFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik J Tokar et al.
Toxicology letters, 209(2), 179-185 (2012-01-11)
Inorganic arsenic, an early life carcinogen in humans and mice, can initiate lesions promotable by other agents in later life. The biomethylation product of arsenic, dimethylarsinic acid (DMA), is a multi-site tumor promoter. Thus, pregnant CD1 mice were given drinking
S Yamamoto et al.
Mutation research, 386(3), 353-361 (1997-06-01)
The modifying effects of dimethylarsinic acid (DMA), the major metabolite of ingested arsenicals in most mammals, on chemical carcinogenesis were investigated using rat in vivo models and reviewed here. In a multi-organ bioassay, rats pretreated with 5 carcinogens were administered
Hua Naranmandura et al.
Toxicological sciences : an official journal of the Society of Toxicology, 128(1), 137-146 (2012-04-28)
Arsenic is known to be a human carcinogen as well as one of the most effective drugs for treatment of patients with acute promyelocytic leukemia. The intermediate metabolites of arsenic, monomethylarsonous acid (MMA(III)) and dimethylarsinous acid (DMA(III)), are formed by
Kavitha Marapakala et al.
Biochemistry, 51(5), 944-951 (2012-01-20)
The enzyme As(III) S-adenosylmethionine methyltransferase (EC 2.1.1.137) (ArsM or AS3MT) is found in members of every kingdom, from bacteria to humans. In these enzymes, there are three conserved cysteine residues at positions 72, 174, and 224 in the CmArsM orthologue
Tomohiro Narukawa et al.
Journal of agricultural and food chemistry, 60(4), 1122-1127 (2012-01-10)
A monitoring test for arsenic species in white rice flour was developed and applied to flours made from 20 samples of polished rice collected from locations all over Japan. The arsenic species in white rice flour made from five samples

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service