G003778
GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
application for SPME, suitable for GC
About This Item
technique(s)
gas chromatography (GC): suitable
solid phase microextraction (SPME): suitable
test parameters
sample/matrix: Phenols at 25 ppb in 0.1M phosphate buffer, pH 2, with 25% NaCl, 9 mL in 10 mL screw cap vial
SPME fiber: 60 μm Carbowax (PEG) (57354-U)
extraction: 30 min direct immersion with agitation using CombiPal autosampler
desorption process: (30 min. direct immersion with agitation using CombiPal autosampler)
column: SLB-5ms, 30 m x 0.25 mm I.D., 0.5μm film
oven: 50°C(2.0 min) to 160°C @ 10°C/min to 280°C @ 20°C/min
inj. temp.: Splitless/split with 0.75mm ID liner and Merlin Microseal
detector: ITMS m/z=45-275 @ 0.7 μsec per scan, transfer line 310°C
carrier gas: Helium, 1mL/min constant flow, 8.7 psi
suitability
application for SPME
application(s)
chemicals and industrial polymers
environmental
food and beverages
Legal Information
Analyte
SPME fiber
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service